missing translation for 'onlineSavingsMsg'
Learn More

Meta-Topolin, MedChemExpress

Product Code. 16495416
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495416 100 mg 100mg
16441606 500 mg 500mg
16400877 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16495416 Supplier Medchem Express Supplier No. HY112104

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Meta-Topolin (m-Topolin) is a highly active aromatic cytokinin.

Meta-Topolin (m-Topolin) is a highly active aromatic cytokinin.

Specifications

Chemical Name or Material Meta-Topolin
CAS 75737-38-1
Color White
Purity Grade Notes Research
Molecular Formula C12H11N5O
Quantity 100 mg
Synonym m-Topolin
Solubility Information DMSO : 125 mg/mL (518.13 mM; Need ultrasonic)
SMILES OC1=CC=CC(CNC2=C3N=CNC3=NC=N2)=C1
Molecular Weight (g/mol) 241.25
Formula Weight 241.25
Percent Purity 98.76%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.