missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meta-Topolin, MedChemExpress
Description
Meta-Topolin (m-Topolin) is a highly active aromatic cytokinin.
Specifications
Specifications
| Chemical Name or Material | Meta-Topolin |
| CAS | 75737-38-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C12H11N5O |
| Quantity | 50 mg |
| Synonym | m-Topolin |
| Solubility Information | DMSO : 125 mg/mL (518.13 mM; Need ultrasonic) |
| SMILES | OC1=CC=CC(CNC2=C3N=CNC3=NC=N2)=C1 |
| Molecular Weight (g/mol) | 241.25 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?