missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Packaging | Ampoule |
| CAS | 2642-71-9 |
| Physical Form | Liquid |
| Molecular Formula | C12H16N3O3PS2 |
| Concentration | 10 μg/mL |
| UN Number | 1145 |
| InChI Key | RQVGAIADHNPSME-UHFFFAOYSA-N |
| Type | Organic Single-component Standard Solution |
| SMILES | CCOP(=S)(OCC)SCN1C(=O)C2=CC=CC=C2N=N1 |
| IUPAC Name | 3-(diethoxyphosphinothioylsulfanylmethyl)-1,2,3-benzotriazin-4-one |
| Show More |
Safety and Handling
Product Identifier
- Standard solution Azinphos-ethyl 10ug/ml in cyclohexane
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Aspiration toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P321-Specific treatment (see on this label).
- P331-Do NOT induce vomiting.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P391-Collect spillage.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?