Learn More
Brady™ MetaLabel™ Series Metallic, Non-Metal Labels, B-413
Product Code. 11529423
Click to view available options
Size (Metric):
101.6 x 50.8 mm (W x H)
12.7 x 11.09 mm (W x H)
19.05 x 6.35 mm (W x H)
25.4 x 12.7 mm (W x H)
25.4 x 9.52 mm (W x H)
38.1 x 12.7 mm (W x H)
38.1 x 19.05 mm (W x H)
50.8 x 12.7 mm (W x H)
50.8 x 25.4 mm (W x H)
76.2 x 25.4 mm (W x H)
76.2 x 50.8 mm (W x H)
W x H: 101.6 mm x 91.4m
W x H: 50.8 mm x 91.4m
W x H: 69.85 mm x 31.75 mm
W x H: 76.2 mm x 91.4m
Unit Size:
1 roll
Product Code. 11529423
Brand: Brady™ 104174
Only null left
Add to basket
Add to basket
Used to identify property from components to finished products. Brady™ MetaLabel™ Series Metallic, Non-Metal Labels are made of Metallic, Non-Metal Polyester.
  • Available in various sizes
  • Acrylic-based, industrial-strength adhesive for multiple applications
Specifications
Certifications/Compliance UL Recognised; cUL Recognised
Description Thermal Transfer Printable Labels
Adhesive Type Acrylic, Permanent
Coating None
Color Light Gray
Finish Satin
Format Roll
Material Polyester (B-480)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Asset Tracking, General and Industrial Identificationentification, Rating Plates
For Use With (Equipment) BBP81, i5100, i6100, i7100, i7500, PR Plus, IP Series, THT 76 mm Core Printers
Labels per Row 1
Min. Operating Temperature (Metric) -70°C
Model THT-53-413-3
No. per Pack 3000 Label(s) / Roll
Quantity 3000/Pk.
Shape Rectangle
Size 50.80 mm (W) x 12.70 mm (H)
Size (Metric) 50.8 x 12.7 mm (W x H)
Temperature Range (English) -94°F to +230°F
Temperature Range (Metric) -70°C to +110°C
Thickness (English) 0.0034 in.
Thickness (Metric) 0.086 mm
Vertical Repeat (Metric) 15.24 mm
Dimensions (LxW) 12.70 x 50.80 mm
Length (English) 0.5 in.
Width (English) 2 in.
Width (Metric) 50.8 mm
Width (English) Printable Area 50.80 mm
Height (English) 0.5 in.
Height (Metric) 12.7 mm
Alternate Part Number THT-53-413-3