Learn More
Brady™ MetaLabel™ Series Metallic, Non-Metal Labels, B-480
Used to identify property from components to finished products. Brady™ MetaLabel™ Series Metallic, Non-Metal Labels are made of Metallic, Non-Metal Polyester.
€ 331.19 - € 1068.55
Specifications
| Coating | None |
|---|---|
| Color | Light Gray |
| Print Technology | Thermal Transfer |
| Product Type | Label |
| For Use With (Application) | Asset Tracking, General and Industrial Identificationentification, Rating Plates |
| Product Code | Brand | Size (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Size (Metric) | Price | Quantity & Availability | |||||
|
11539443
|
Brady™
104196 |
W x H: 50.8 mm x 91.4m |
€ 575.00 € 523.34 / 1 rolls Save € 51.66 8% Off |
Estimated Shipment: 17-12-2026
Log in to see stock. |
|||||
|
11549443
|
Brady™
104197 |
W x H: 101.6 mm x 91.4m |
€ 1160.00 € 1068.55 / 1 rolls Save € 91.45 7% Off |
In Stock
Log in to see stock. |
|||||
|
11519433
|
Brady™
104183 |
19.05 x 6.35 mm (W x H) |
€ 468.00 € 432.85 / 1 rolls Save € 35.15 7% Off |
In Stock
Log in to see stock. |
|||||
|
11579433
|
Brady™
104190 |
50.8 x 25.4 mm (W x H) |
€ 390.00 € 360.40 / 1 rolls Save € 29.60 7% Off |
In Stock
Log in to see stock. |
|||||
|
11599433
|
Brady™
104192 |
W x H: 76.2 mm x 25.4 mm |
€ 560.00 € 511.27 / 1 rolls Save € 48.73 8% Off |
In Stock
Log in to see stock. |
|||||
|
11509443
|
Brady™
104193 |
W x H: 76.2 mm x 50.8 mm |
€ 369.00 € 340.79 / 1 rolls Save € 28.21 7% Off |
In Stock
Log in to see stock. |
|||||
|
11529443
|
Brady™
104195 |
W x H: 76.2 mm x 91.4m |
€ 790.00 € 727.41 / 1 rolls Save € 62.59 7% Off |
In Stock
Log in to see stock. |
|||||
|
11529433
|
Brady™
104184 |
25.4 x 9.52 mm (W x H) |
€ 560.00 € 513.01 / 1 rolls Save € 46.99 8% Off |
In Stock
Log in to see stock. |
|||||
|
11559433
|
Brady™
104187 |
38.1 x 12.7 mm (W x H) |
€ 995.00 € 916.24 / 1 rolls Save € 78.76 7% Off |
In Stock
Log in to see stock. |
|||||
|
11569433
|
Brady™
104189 |
50.8 mm x 12.7 mm (W x H) |
€ 358.00 € 331.19 / 1 rolls Save € 26.81 7% Off |
In Stock
Log in to see stock. |
|||||
|
11519443
|
Brady™
104194 |
W x H: 101.6 mm x 50.8 mm |
€ 467.00 € 421.43 / 1 rolls Save € 45.57 9% Off |
In Stock
Log in to see stock. |
|||||
|
11549433
|
Brady™
104186 |
38.1 x 19.05 mm (W x H) |
€ 1045.00 € 962.09 / 1 rolls Save € 82.91 7% Off |
In Stock
Log in to see stock. |
|||||
|
11589433
|
Brady™
104191 |
W x H: 69.85 mm x 31.75 mm |
€ 635.00 € 581.07 / 1 rolls Save € 53.93 8% Off |
In Stock
Log in to see stock. |
|||||
Description
- Available in various sizes
- Rubber-based Mondo Bondo™ adhesive for oily, greasy applications
Specifications
| None | |
| Thermal Transfer | |
| Asset Tracking, General and Industrial Identificationentification, Rating Plates | |
| Rectangle | |
| -70°C to +110°C | |
| 0.112 mm |
| Light Gray | |
| Label | |
| -70°C | |
| -94°F to +230°F | |
| 0.0044 in. |