missing translation for 'onlineSavingsMsg'
Learn More
Learn More
metallothionein Antibody (2B5), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18262UV
This item is not returnable.
View return policy
Description
metallothionein Monoclonal antibody specifically detects metallothionein in Bacteria samples. It is validated for Western Blot, ELISA
Specifications
| metallothionein | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Recombinant full length Pseudomonas aeruginosa Metallothionein protein | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| 2B5 | |
| Western Blot, ELISA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction