missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metapneumovirus Antibody (hMPV57), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53105AF488
This item is not returnable.
View return policy
Description
Metapneumovirus Monoclonal antibody specifically detects Metapneumovirus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Metapneumovirus | |
| Monoclonal | |
| Alexa Fluor 488 | |
| hMPV | |
| Human MPV strain NCL-174 | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| hMPV57 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction