missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metapneumovirus Fusion Protein Antibody (hMPV24) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50505
This item is not returnable.
View return policy
Description
Metapneumovirus Fusion Protein Monoclonal antibody specifically detects Metapneumovirus Fusion Protein in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Metapneumovirus Fusion Protein | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 20 μg/mL | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| hMPV24 | |
| Unconjugated | |
| PBS | |
| Human MPV strain NCL-145 | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction