missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab11A Antibody (STo 7C10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50260H
This item is not returnable.
View return policy
Description
Rab11A Monoclonal antibody specifically detects Rab11A in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| Rab11A | |
| Monoclonal | |
| HRP | |
| MGC1490, RAB 11A, member oncogene family, Rab-11, RAB11, RAB11A, member RAS oncogene family, YL8ras-related protein Rab-11A | |
| Rab11A peptide KQMSDRRENDM | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 8766 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunoprecipitation | |
| STo 7C10 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction