missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metapneumovirus Fusion Protein Antibody (hMPV24), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50505F
This item is not returnable.
View return policy
Description
Metapneumovirus Fusion Protein Monoclonal specifically detects Metapneumovirus Fusion Protein in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Metapneumovirus Fusion Protein | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Human MPV strain NCL-145 | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| Virus | |
| Purified |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| hMPV24 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction