missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Metapneumovirus matrix Monoclonal Antibody (D87K)
GREENER_CHOICE

Product Code. 15392697 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15392697 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15392697 Supplier Invitrogen™ Supplier No. MA518225

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This antibody is specific for the matrix protein of human metapneumovirus. It does not cross-react with Respiratory Syncytial Virus, Influenza A, Influenza B, Adenovirus or Parainfluenza viruses 1, 2 & 3. Purity is >90%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Metapneumovirus matrix
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone D87K
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias hMPV; h-MPV; hMPV matrix; MPV matrix protein
Host Species Mouse
Immunogen Metapneumovirus recombinant protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.