missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metapneumovirus Phosphoprotein Antibody (hMPV33), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50506AF750
This item is not returnable.
View return policy
Description
Metapneumovirus Phosphoprotein Monoclonal specifically detects Metapneumovirus Phosphoprotein in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Metapneumovirus Phosphoprotein | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| Human hMPV strain NCL-174 | |
| 0.1 mL | |
| Primary | |
| Virus | |
| IgG2a |
| ELISA, Immunocytochemistry, Immunofluorescence | |
| hMPV33 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| Virology Bacteria and Parasites | |
| Identifies Metapneumovirus nucleoprotein of both types A and B. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction