missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Meteorin Monoclonal Antibody (347505)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA524084
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Meteorin is a gene encoding a protein that is part of the cytokine and growth factor family, primarily involved in neurogenesis and gliogenesis. It is located on chromosome 17p13.1 and is highly expressed in the nervous system, where it plays a role in promoting the differentiation and maturation of astrocytes and neural precursor cells. Meteorin functions by enhancing neurite outgrowth and synaptic development, crucial for proper neural network formation. Additionally, it has been implicated in angiogenesis, contributing to the development and maintenance of blood vessels in neural tissue. Research suggests that Meteorin has neuroprotective properties and may help in recovery from neural injury, making it a molecule of interest for therapeutic strategies in neurodegenerative diseases and brain injuries. Its expression and activity are also being studied in relation to inflammatory responses and potential roles in other tissues, offering insights into its diverse biological functions beyond the central nervous system.
Specifications
| Meteorin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8C1Q4 | |
| METRN | |
| Mouse myeloma cell line NS0-derived recombinant mouse Meteorin isoform 1 Gly22-Asp291. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| 347505 | |
| Unconjugated | |
| METRN | |
| 1810034B16Rik; C16orf23; c380A1.2; hypoxia/reoxygenation regulatory factor; hypoxia/reoxygenation regulatory factor in astrocytes; Hyrac; Meteorin; meteorin, glial cell differentiation regulator; METRN; RGD1306222; RJD6 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 70083 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction