missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Methadone Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA175265
This item is not returnable.
View return policy
Description
Working Concentration: 1/32K. Sensitivity: 10 ng/mL Methadone produces 79% inhibition in a competitive ELISA employing Methadone polyclonal antibody.
Methadone is a synthetic opioid which is used widely as an analgesic as well as maintenance therapy for persons with opioid dependency. Patients on chronic methadone therapy for opioid dependency often have underlying chronic viral hepatitis, but methadone itself has not been linked to serum enzyme elevations during therapy or to clinically apparent liver injury. [National Center for Biotechnology Information. PubChem Compound Database; CID=4095].
Specifications
| Methadone | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.8 | |
| Sheep | |
| Purified | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
| ELISA | |
| Unconjugated | |
| Amidone; C21H27NO; Dl-Methadone; Dolophine; Methadol; Phenadone; Physeptone; Racemethadol | |
| Methadone-BSA. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction