missing translation for 'onlineSavingsMsg'
Learn More

Methanesulfonic Acid 3-(3-Trifluoromethylphenyl)propyl Ester, TRC

Product Code. 17905095
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17905095 50 mg 50mg
17904925 25 mg 25mg
17991005 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17905095 Supplier Toronto Research Chemicals Supplier No. TRCM22578050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21172-43-0
Molecular Formula C11 H13 F3 O3 S
Molecular Weight (g/mol) 282.28
Synonym Benzenepropanol, 3-(trifluoromethyl)-, 1-methanesulfonate, 1-Propanol, 3-(α,α,α-trifluoro-m-tolyl)-, methanesulfonate (8CI), Benzenepropanol, 3-(trifluoromethyl)-, methanesulfonate (9CI), 3-(3-Trifluoromethylphenyl)propyl methanesulfonate, Methanesulfonic acid 3-(3-trifluoromethylphenyl)propyl ester, 3-(trifluoromethyl)Benzenepropanal, Methanesulfonate, 1-Propanol, 3-(α,α,α-trifluoro-m-tolyl)-, Methanesulfonate, Benzenepropanol, 3-(trifluoromethyl)-, 1-Methanesulfonate
IUPAC Name 3-[3-(trifluoromethyl)phenyl]propyl methanesulfonate
SMILES CS(=O)(=O)OCCCc1cccc(c1)C(F)(F)F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C11H13F3O3S/c1-18(15,16)17-7-3-5-9-4-2-6-10(8-9)11(12,13)14/h2,4,6,8H,3,5,7H2,1H3
Formula Weight 282.0537
Chemical Name or Material Methanesulfonic Acid 3-(3-Trifluoromethylphenyl)propyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.