Learn More
Methanesulfonic Acid 3-(3-Trifluoromethylphenyl)propyl Ester, TRC
Product Code. 17991005
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991005 100 mg 100mg
17905095 50 mg 50mg
17904925 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17991005 Supplier Toronto Research Chemicals Supplier No. TRCM225780100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21172-43-0
Molecular Formula C11 H13 F3 O3 S
Molecular Weight (g/mol) 282.28
Synonym Benzenepropanol, 3-(trifluoromethyl)-, 1-methanesulfonate, 1-Propanol, 3-(α,α,α-trifluoro-m-tolyl)-, methanesulfonate (8CI), Benzenepropanol, 3-(trifluoromethyl)-, methanesulfonate (9CI), 3-(3-Trifluoromethylphenyl)propyl methanesulfonate, Methanesulfonic acid 3-(3-trifluoromethylphenyl)propyl ester, 3-(trifluoromethyl)Benzenepropanal, Methanesulfonate, 1-Propanol, 3-(α,α,α-trifluoro-m-tolyl)-, Methanesulfonate, Benzenepropanol, 3-(trifluoromethyl)-, 1-Methanesulfonate
IUPAC Name 3-[3-(trifluoromethyl)phenyl]propyl methanesulfonate
SMILES CS(=O)(=O)OCCCc1cccc(c1)C(F)(F)F
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C11H13F3O3S/c1-18(15,16)17-7-3-5-9-4-2-6-10(8-9)11(12,13)14/h2,4,6,8H,3,5,7H2,1H3
Formula Weight 282.0537
Chemical Name or Material Methanesulfonic Acid 3-(3-Trifluoromethylphenyl)propyl Ester