missing translation for 'onlineSavingsMsg'
Learn More

Methapyrilene Hydrochloride, TRC

Product Code. 30477587
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477587 250 mg 250mg
30475110 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30477587 Supplier Toronto Research Chemicals Supplier No. TRCM259970250MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-23-9
Molecular Formula C14 H19 N3 S . Cl H
Molecular Weight (g/mol) 297.85
Synonym Thenylpyramine Hydrochloride, 1,2-Ethanediamine, N,N-dimethyl-N'-2-pyridinyl-N'-(2-thienylmethyl)-, monohydrochloride, Pyridine, 2-[[2-(dimethylamino)ethyl]-2-thenylamino]-, monohydrochloride, 2-[[2-(Dimethylamino)ethyl]-2-thenylamino]pyridine hydrochloride, Barhist, Blue Line, Bu-pyra-hist, Bu-pyra-histrit, Butyl-pyra-hist, Capathyn, Cohistine, Coryzol, Histadyl, Histafed, Lullamin, Methacon, Methapyrilene hydrochloride, Methoxylene, N,N-Dimethyl-N'-(2-thenyl)-N'-(2-pyridyl-ethylene-diamine hydrochloride), Pyrathyn, Somnicaps, Tem-Histine, Teralin, Thenylene hydrochloride
IUPAC Name N,N-dimethyl-N'-pyridin-2-yl-N'-(thiophen-2-ylmethyl)ethane-1,2-diamine;hydrochloride
SMILES Cl.CN(C)CCN(Cc1cccs1)c2ccccn2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H19N3S.ClH/c1-16(2)9-10-17(12-13-6-5-11-18-13)14-7-3-4-8-15-14;/h3-8,11H,9-10,12H2,1-2H3;1H
Formula Weight 297.1066
Percent Purity >95%
Chemical Name or Material Methapyrilene hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.