Learn More
Methenolone Acetate, Mikromol™
Description
Methenolone Acetate
Specifications
Specifications
| Chemical Name or Material | Methenolone Acetate |
| CAS | 434-05-9 |
| Analyte or Component Names | Methenolone Acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H32 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Androst-1-en-3-one, 17-(acetyloxy)-1-methyl-, (5α,17β)-, Methenolone Acetate, 5α-Androst-1-en-3-one, 17β-hydroxy-1-methyl-, acetate (6CI,8CI), (5α,17β)-17-(Acetyloxy)-1-methylandrost-1-en-3-one, 1-Methyl-17β-hydroxy-5α-androst-1-en-3-one acetate, 1-Methyl-17β-hydroxy-5α-androst-1-en-3-one-17β-acetate, 1-Methyl-5α-androst-1-en-17β-ol-3-one acetate, 17β-Hydroxy-1-methyl-5α-androst-1-en-3-one acetate, Metenolone acetate, Methenolone 17-acetate, Methenolone acetate, NSC 74226, Nibal, Primobolan, Primobolan Tablets, Primobolone, Primonabol, SH 567, SH 567a, SQ 16496 |
| InChI Formula | InChI=1S/C22H32O3/c1-13-11-16(24)12-15-5-6-17-18-7-8-20(25-14(2)23)21(18,3)10-9-19(17)22(13,15)4/h11,15,17-20H,5-10,12H2,1-4H3/t15-,17-,18-,19-,20-,21-,22-/m0/s1 |
| SMILES | CC(=O)O[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CC(=O)C=C(C)[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | [(5S,8R,9S,10S,13S,14S,17S)-1,10,13-trimethyl-3-oxo-4,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate |
| Show More |