missing translation for 'onlineSavingsMsg'
Learn More

Methenolone Acetate, TRC

Product Code. 30470579
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470579 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30470579 Supplier Toronto Research Chemicals Supplier No. TRCM2262501G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methenolone Acetate
CAS 434-05-9
Molecular Formula C22 H32 O3
Quantity 1 g
Synonym Androst-1-en-3-one, 17-(acetyloxy)-1-methyl-, (5α,17β)-, Methenolone Acetate, 5α-Androst-1-en-3-one, 17β-hydroxy-1-methyl-, acetate (6CI,8CI), (5α,17β)-17-(Acetyloxy)-1-methylandrost-1-en-3-one, 1-Methyl-17β-hydroxy-5α-androst-1-en-3-one acetate, 1-Methyl-17β-hydroxy-5α-androst-1-en-3-one-17β-acetate, 1-Methyl-5α-androst-1-en-17β-ol-3-one acetate, 17β-Hydroxy-1-methyl-5α-androst-1-en-3-one acetate, Metenolone acetate, Methenolone 17-acetate, Methenolone acetate, NSC 74226, Nibal, Primobolan, Primobolan Tablets, Primobolone, Primonabol, SH 567, SH 567a, SQ 16496
InChI Formula InChI=1S/C22H32O3/c1-13-11-16(24)12-15-5-6-17-18-7-8-20(25-14(2)23)21(18,3)10-9-19(17)22(13,15)4/h11,15,17-20H,5-10,12H2,1-4H3/t15-,17-,18-,19-,20-,21-,22-/m0/s1
SMILES CC(=O)O[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CC(=O)C=C(C)[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(5S,8R,9S,10S,13S,14S,17S)-1,10,13-trimethyl-3-oxo-4,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 344.49
Formula Weight 344.2351
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.