missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methiocarb-sulfoxide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Methiocarb sulfoxide |
| CAS | 2635-10-1 |
| Analyte or Component Names | Methiocarb sulfoxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H15 N O3 S |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Carbamic acid, methyl-, 4-(methylsulfinyl)-3,5-xylyl ester (7CI,8CI), Phenol, 3,5-dimethyl-4-(methylsulfinyl)-, methylcarbamate (9CI), 3,5-Xylenol, 4-(methylsulfinyl)-, methylcarbamate (8CI), 4-(Methylsulfinyl)-3,5-xylyl methylcarbamate, BAY 41791, Bay 37344 sulfoxide, Mesurol sulfoxide, Methiocarb sulfoxide, Methiocarb-sulfoxide |
| InChI Formula | InChI=1S/C11H15NO3S/c1-7-5-9(15-11(13)12-3)6-8(2)10(7)16(4)14/h5-6H,1-4H3,(H,12,13) |
| SMILES | CNC(=O)Oc1cc(C)c(c(C)c1)S(=O)C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?