missing translation for 'onlineSavingsMsg'
Learn More

Methiocarb Sulfoxide, TRC

Product Code. 30464708
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464708 2.5 g 2.5g
30476103 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30464708 Supplier Toronto Research Chemicals Supplier No. TRCM2603502.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2635-10-1
Molecular Formula C11 H15 N O3 S
Molecular Weight (g/mol) 241.31
Synonym Carbamic acid, methyl-, 4-(methylsulfinyl)-3,5-xylyl ester (7CI,8CI), Phenol, 3,5-dimethyl-4-(methylsulfinyl)-, methylcarbamate (9CI), 3,5-Xylenol, 4-(methylsulfinyl)-, methylcarbamate (8CI), 4-(Methylsulfinyl)-3,5-xylyl methylcarbamate, BAY 41791, Bay 37344 sulfoxide, Mesurol sulfoxide, Methiocarb sulfoxide, Methiocarb-sulfoxide
IUPAC Name (3,5-dimethyl-4-methylsulfinylphenyl) N-methylcarbamate
SMILES CNC(=O)Oc1cc(C)c(c(C)c1)S(=O)C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C11H15NO3S/c1-7-5-9(15-11(13)12-3)6-8(2)10(7)16(4)14/h5-6H,1-4H3,(H,12,13)
Formula Weight 241.0773
Chemical Name or Material Methiocarb sulfoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.