missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methionine Aminopeptidase 1D/MAP1D Antibody (008), DyLight 755 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89693IR
This item is not returnable.
View return policy
Description
Methionine Aminopeptidase 1D/MAP1D Monoclonal antibody specifically detects Methionine Aminopeptidase 1D/MAP1D in Human samples. It is validated for ELISA
Specifications
| Methionine Aminopeptidase 1D/MAP1D | |
| Monoclonal | |
| DyLight 755 | |
| EC 3.4.11.18, MAP1DCDS of metAP-3 within PCR fragment, Metap1l, methionine aminopeptidase 1D, mitochondrial, methionyl aminopeptidase type 1D (mitochondrial), Methionyl aminopeptidase type 1D, mitochondrial | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Methionine Aminopeptidase 1D/MAP1D (NP_954697.1; Arg 44-Ala 335). | |
| 0.1 mL | |
| Immunology | |
| 254042 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 008 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction