missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Methionyl tRNA synthetase Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22313PCP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Methionyl tRNA synthetase Polyclonal antibody specifically detects Methionyl tRNA synthetase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifikationer
| Methionyl tRNA synthetase | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| cytosolic methionyl-tRNA synthetase, EC 6.1.1.10, FLJ35667, methionine tRNA ligase 1, cytoplasmic, Methionine--tRNA ligase, methionyl-tRNA synthetase, methionyl-tRNA synthetase, cytoplasmic, METRS, MRS, MTRNS | |
| The immunogen this antibody was made to, maps to a region between residue 650 to 700 of human Methionyl-tRNA Synthetase using the numbering given in entry NP_004981.2 (GeneID 4141). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 4141 | |
| Store at 4C in the dark. | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering