missing translation for 'onlineSavingsMsg'
Learn More

Methisosildenafil, TRC

Product Code. 30138460
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138460 5 mg 5mg
30136820 10 mg 10mg
30006043 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138460 Supplier Toronto Research Chemicals Supplier No. TRCM2259355MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496835-35-9
Molecular Formula C23 H32 N6 O4 S
Molecular Weight (g/mol) 488.6
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[5-[[(3R,5S)-3,5-dimethyl-1-piperazinyl]sulfonyl]-2-ethoxyphenyl]-1,6-dihydro-1-methyl-3-propyl-, rel-, Piperazine, 1-[[3-(4,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-3,5-dimethyl-, (3R,5S)-rel- (9CI), Aildenafil, Methisosildenafil
IUPAC Name 5-[5-[(3S,5R)-3,5-dimethylpiperazin-1-yl]sulfonyl-2-ethoxyphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4C[C@@H](C)N[C@@H](C)C4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C23H32N6O4S/c1-6-8-18-20-21(28(5)27-18)23(30)26-22(25-20)17-11-16(9-10-19(17)33-7-2)34(31,32)29-12-14(3)24-15(4)13-29/h9-11,14-15,24H,6-8,12-13H2,1-5H3,(H,25,26,30)/t14-,15+
Formula Weight 488.2206
Percent Purity >95%
Chemical Name or Material Methisosildenafil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.