missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies MethoCult™ H4230
Click to view available options
Quantity:
80 mL
Unit Size:
80mL
Description
- MethoCult™ H4230 is an incomplete methylcellulose-based medium for the growth and enumeration of hematopoietic progenitor cells in colony-forming unit (CFU) assays of human bone marrow, mobilized peripheral blood, peripheral blood, and cord blood samples
- This formulation does not contain erythropoietin (EPO) or other cytokines, allowing the addition of cytokines to meet the specific requirements of investigators
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Functional Assay |
| Quantity | 80 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction