missing translation for 'onlineSavingsMsg'
Learn More

Methopterine (>90%), TRC

Product Code. 30135930
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135930 1 g 1g
30012871 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135930 Supplier Toronto Research Chemicals Supplier No. TRCM3053701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2410-93-7
Molecular Formula C20 H21 N7 O6
Molecular Weight (g/mol) 455.42
Synonym N-[4-[[(2-Amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]methylamino]benzoyl]-L-glutamic Acid, Glutamic acid, N-[p-[[(2-amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]methylamino]benzoyl]-, L- (7CI,8CI), Glutamic acid, N-[p-[[(2-amino-4-hydroxy-6-pteridinyl)methyl]methylamino]benzoyl]- (6CI), L-Glutamic acid, N-[4-[[(2-amino-1,4-dihydro-4-oxo-6-pteridinyl)methyl]methylamino]benzoyl]- (9CI), 10-Methylfolic acid, 10-Methylpteroylglutamic acid, Methopterin, Methopterine, N10-Methylfolic acid, N10-Methylpteroylglutamic acid, NSC 107144
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
SMILES CN(Cc1cnc2NC(=NC(=O)c2n1)N)c3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C20H21N7O6/c1-27(9-11-8-22-16-15(23-11)18(31)26-20(21)25-16)12-4-2-10(3-5-12)17(30)24-13(19(32)33)6-7-14(28)29/h2-5,8,13H,6-7,9H2,1H3,(H,24,30)(H,28,29)(H,32,33)(H3,21,22,25,26,31)/t13-/m0/s1
Formula Weight 455.1553
Chemical Name or Material Methopteridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.