missing translation for 'onlineSavingsMsg'
Learn More

Methotrexate, TRC

Product Code. 30470116
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30470116

Brand: Toronto Research Chemicals TRCM2606751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methotrexate
CAS 59-05-2
Purity Grade Notes HPLC
Molecular Formula C20 H22 N8 O5
Quantity 1 g
Synonym N-[4-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-L-glutamic acid, L-(+)-N-[p-[[(2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-glutamic acid, (+)-Amethopterin, 4-Amino-10-methylfolic acid, 4-Amino-N10-methylfolic acid, 4-Amino-N10-methylpteroylglutamic acid, Amethopterin, Amethopterine, Antifolan, CL 14377, EMT 25299, Emtexate, L-Amethopterin, L-Methotrexate, Ledertrexate, MTX, Metatrexan, Methotrexat-Ebewe, Methotrexate, Methoxtrexate, Methylaminopterin, Mexate, N-[p-[[2,4-Diamino-6-pteridinyl)methyl]methylamino]benzoyl]-L-(+)-glutamic acid, NSC 740, R 9985, Rheumatrex, Trexall
InChI Formula InChI=1S/C20H22N8O5/c1-28(9-11-8-23-17-15(24-11)16(21)26-20(22)27-17)12-4-2-10(3-5-12)18(31)25-13(19(32)33)6-7-14(29)30/h2-5,8,13H,6-7,9H2,1H3,(H,25,31)(H,29,30)(H,32,33)(H4,21,22,23,26,27)/t13-/m0/s1
SMILES CN(Cc1cnc2nc(N)nc(N)c2n1)c3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O
IUPAC Name (2S)-2-[[4-[(2,4-diaminopteridin-6-yl)methyl-methylamino]benzoyl]amino]pentanedioic acid
Molecular Weight (g/mol) 454.44
Formula Weight 454.1713
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.