missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methoxy Empagliflozin Tetraacetate, TRC
Specifications
Specifications
| Chemical Name or Material | Empagliflozin Impurity 20 |
| CAS | 872980-38-6 |
| Molecular Formula | C28 H31 Cl O10 |
| Quantity | 100 mg |
| Synonym | D-Glucitol, 1,5-anhydro-1-C-[4-chloro-3-[(4-methoxyphenyl)methyl]phenyl]-, 2,3,4,6-tetraacetate, (1S)-, D-Glucitol, 1,5-anhydro-1-C-[4-chloro-3-[(4-methoxyphenyl)methyl]phenyl]-, tetraacetate, (1S)- (9CI), [(2R,3R,4R,5S,6S)-3,4,5-triacetoxy-6-[4-chloro-3-[(4-methoxyphenyl)methyl]phenyl]tetrahydropyran-2-yl]methyl acetate |
| InChI Formula | InChI=1S/C28H31ClO10/c1-15(30)35-14-24-26(36-16(2)31)28(38-18(4)33)27(37-17(3)32)25(39-24)20-8-11-23(29)21(13-20)12-19-6-9-22(34-5)10-7-19/h6-11,13,24-28H,12,14H2,1-5H3/t24-,25+,26-,27+,28+/m1/s1 |
| SMILES | COc1ccc(Cc2cc(ccc2Cl)[C@@H]3O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]3OC(=O)C)cc1 |
| IUPAC Name | [(2R,3R,4R,5S,6S)-3,4,5-triacetyloxy-6-[4-chloro-3-[(4-methoxyphenyl)methyl]phenyl]oxan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 562.993 |
| Formula Weight | 562.161 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?