missing translation for 'onlineSavingsMsg'
Learn More

Methoxy Empagliflozin Tetraacetate, TRC

Product Code. 30379343
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379343 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30379343 Supplier Toronto Research Chemicals Supplier No. TRCE87298010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Empagliflozin Impurity 20
CAS 872980-38-6
Molecular Formula C28 H31 Cl O10
Quantity 10 mg
Synonym D-Glucitol, 1,5-anhydro-1-C-[4-chloro-3-[(4-methoxyphenyl)methyl]phenyl]-, 2,3,4,6-tetraacetate, (1S)-, D-Glucitol, 1,5-anhydro-1-C-[4-chloro-3-[(4-methoxyphenyl)methyl]phenyl]-, tetraacetate, (1S)- (9CI), [(2R,3R,4R,5S,6S)-3,4,5-triacetoxy-6-[4-chloro-3-[(4-methoxyphenyl)methyl]phenyl]tetrahydropyran-2-yl]methyl acetate
InChI Formula InChI=1S/C28H31ClO10/c1-15(30)35-14-24-26(36-16(2)31)28(38-18(4)33)27(37-17(3)32)25(39-24)20-8-11-23(29)21(13-20)12-19-6-9-22(34-5)10-7-19/h6-11,13,24-28H,12,14H2,1-5H3/t24-,25+,26-,27+,28+/m1/s1
SMILES COc1ccc(Cc2cc(ccc2Cl)[C@@H]3O[C@H](COC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@H]3OC(=O)C)cc1
IUPAC Name [(2R,3R,4R,5S,6S)-3,4,5-triacetyloxy-6-[4-chloro-3-[(4-methoxyphenyl)methyl]phenyl]oxan-2-yl]methyl acetate
Molecular Weight (g/mol) 562.993
Formula Weight 562.161
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.