Learn More
Methoxyfenozide-d9 (3,5-dimethylbenzoyl-d9), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Methoxyfenozide-d9 |
| CAS | 2469014-53-5 |
| Molecular Formula | C22 D9 H19 N2 O3 |
| Quantity | 0.01 g |
| Synonym | Methoxyfenozide D9 (Dimethylbenzoyl D9), Methoxyfenozide-d9, 3-Methoxy-2-methylbenzoic acid 2-[3,5-di(methyl-d3)benzoyl-2,4,6-d3]-2-(1,1-dimethylethyl)hydrazide, 3,5-Di(methyl-d3)benzoic-2,4,6-d3 acid N-tert-butyl-N-(3-methoxy-2-methylbenzoyl)hydrazide |
| InChI Formula | InChI=1 S/C22H28N2O3/c1-14-11-15(2)13-17(12-14)21(26)24(22(4,5)6)23-20(25)18-9-8-10-19(27-7)16(18)3/h8-13 H,1-7H3,(H,23,25)/i1D3,2D3,11 D,12 D,13 D |
| SMILES | [2 H]c1c(C(=O)N(NC(=O)c2cccc(OC)c2C)C(C)(C)C)c([2 H])c(c([2 H])c1C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | N-tert-butyl-2,4,6-trideuterio-N'-(3-methoxy-2-methylbenzoyl)-3,5-bis(trideuteriomethyl)benzohydrazide |
| Molecular Weight (g/mol) | 377.5248 |
| Formula Weight | 377.2665 g/mol |
| Show More |