missing translation for 'onlineSavingsMsg'
Learn More

Methyl 1-aminocyclobutanecarboxylate hydrochloride, 95%, Thermo Scientific™

Catalog Number 15439086
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
92398-47-5
C6H11NO2
129.16
MFCD01318265
SGXOPTPGZBLQAY-UHFFFAOYSA-N
methyl 1-aminocyclobutanecarboxylate, 1-amino-cyclobutanecarboxylic acid methyl ester, 1-aminocyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, 1-amino-, methyl ester, 1-amino-cyclobutanecarboxylicacidmethylester, methyl 1-amino-cyclobutane-1-carboxylate, 1-aminocyclobutane-1-carboxylic acid methyl ester, cyclobutanecarboxylicacid, 1-amino-, methyl ester
1502033
methyl 1-aminocyclobutane-1-carboxylate
COC(=O)C1(N)CCC1
This item is not returnable. View return policy
92398-47-5
C6H11NO2
129.16
MFCD01318265
SGXOPTPGZBLQAY-UHFFFAOYSA-N
methyl 1-aminocyclobutanecarboxylate, 1-amino-cyclobutanecarboxylic acid methyl ester, 1-aminocyclobutanecarboxylic acid methyl ester, cyclobutanecarboxylic acid, 1-amino-, methyl ester, 1-amino-cyclobutanecarboxylicacidmethylester, methyl 1-amino-cyclobutane-1-carboxylate, 1-aminocyclobutane-1-carboxylic acid methyl ester, cyclobutanecarboxylicacid, 1-amino-, methyl ester
1502033
methyl 1-aminocyclobutane-1-carboxylate
COC(=O)C1(N)CCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as a primary and secondary intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 5615121
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 165.62
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Methyl 1-aminocyclobutanecarboxylate hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'