missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2,2,3,3-tetrafluoropropyl carbonate, 99%, Thermo Scientific™

Product Code. 15405407
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15405407

Brand: Thermo Scientific Alfa Aesar H32657.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156783-98-1
Molecular Formula C5H6F4O3
Molecular Weight (g/mol) 190.094
MDL Number MFCD08688580
InChI Key LDTVHHNIXCBROE-UHFFFAOYSA-N
Synonym methyl-2,2,3,3-tetrafluoropropyl carbonate, 2,2,3,3-tetrafluoropropyl methoxyformate, methyl 2,2,3,3-tetrafluoroprop-1-yl carbonat, carbonic acid methyl 2,2,3,3-tetrafluoropropyl ester
PubChem CID 21263425
IUPAC Name methyl 2,2,3,3-tetrafluoropropyl carbonate
SMILES COC(=O)OCC(C(F)F)(F)F

Specifications

Refractive Index 1.345
Quantity 1 g
Formula Weight 190.1
Percent Purity 99%
Chemical Name or Material Methyl 2,2,3,3-tetrafluoropropyl carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.