Learn More
Methyl 2-hydroxy-5-nitrobenzoate, 98%
Product Code. p-7028136
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. 11470848
Brand: Thermo Scientific Alfa Aesar B25278.09
Request Bulk or Custom Format
CAS: 17302-46-4 | C8H7NO5 | 197.146 g/mol
Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 2-hydroxy-5-nitrobenzoate is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17302-46-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00024695
InChI Key UUBFELFUKFJSRD-UHFFFAOYSA-N
Synonym methyl 2-hydroxy-5-nitrobenzoate, methyl 5-nitrosalicylate, methyl-5-nitrosalicylate, 5-nitrosalicylic acid methyl ester, benzoic acid, 2-hydroxy-5-nitro-, methyl ester, 2-hydroxy-5-nitrobenzoic acid methyl ester, methyl 5-nitro-salicylate, acmc-20a62q, methyl 5-nitro-2-hydroxybenzoate, methyl-2-hydroxy-5-nitrobenzoate
PubChem CID 87041
SMILES COC(=O)C1=C(C=CC(=C1)[N+](=O)[O-])O
Specifications
Melting Point 114°C to 117°C
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material Methyl 2-hydroxy-5-nitrobenzoate