missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-methylpropionimidate hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number 15448286
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
39739-60-1
C5H12ClNO
137.61
MFCD03844704
RMXGPHAHPIIESQ-UHFFFAOYSA-N
methyl 2-methylpropanecarboximidate hydrochloride, methyl 2-methylpropanimidate hydrochloride, methyl 2-methylpropanimidic acid hydrochloride, methyl 2-methylpropanimidoate hydrochloride, methyl isobutyrimidate hydrochloride, methyl 2-methylpropanimidate-hydrogen chloride 1/1, methyl 1z-2-methylpropanimidoate hydrochloride 1:1
371709
methyl 2-methylpropanimidate;hydrochloride
Cl.COC(=N)C(C)C
This item is not returnable. View return policy
39739-60-1
C5H12ClNO
137.61
MFCD03844704
RMXGPHAHPIIESQ-UHFFFAOYSA-N
methyl 2-methylpropanecarboximidate hydrochloride, methyl 2-methylpropanimidate hydrochloride, methyl 2-methylpropanimidic acid hydrochloride, methyl 2-methylpropanimidoate hydrochloride, methyl isobutyrimidate hydrochloride, methyl 2-methylpropanimidate-hydrogen chloride 1/1, methyl 1z-2-methylpropanimidoate hydrochloride 1:1
371709
methyl 2-methylpropanimidate;hydrochloride
Cl.COC(=N)C(C)C

Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2-methylpropionimidate hydrochloride, is used as a fine chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Linear Formula (CH3)2CHC(=NH)OCH3·HCl
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 137.61
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 2-methylpropionimidate hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'