Learn More
Methyl (2RS)-2-Cyclopentyl-2-hydroxy-2-phenylacetate, Mikromol™
Description
Methyl (2RS)-2-Cyclopentyl-2-hydroxy-2-phenylacetate
Specifications
Specifications
| Chemical Name or Material | Methyl (2RS)-2-Cyclopentyl-2-hydroxy-2-phenylacetate |
| CAS | 19833-96-6 |
| Analyte or Component Names | Methyl (2RS)-2-Cyclopentyl-2-hydroxy-2-phenylacetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H18 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methyl 2-Cyclopentyl-2-hydroxy-2-phenylacetate, Methyl cyclopentylphenylglycolate, NSC 93811, Glycopyrronium Bromide Imp. L (EP), Glycopyrrolate USP Related Compound L, Glycopyrrolate USP RC L, Benzeneacetic acid, alpha-cyclopentyl-alpha-hydroxy-, methyl ester, Mandelic acid, alpha-cyclopentyl-, methyl ester (8CI), Methyl (+/-)-cyclopentylmandelate, Methyl alpha-cyclopentylmandelate, Glycopyrrolate Related Compound L, Glycopyrronium Bromide Impurity L, Glycopyrronium Impurity L |
| InChI Formula | InChI=1S/C14H18O3/c1-17-13(15)14(16,12-9-5-6-10-12)11-7-3-2-4-8-11/h2-4,7-8,12,16H,5-6,9-10H2,1H3 |
| SMILES | COC(=O)C(O)(C1CCCC1)c2ccccc2 |
| IUPAC Name | methyl 2-cyclopentyl-2-hydroxy-2-phenylacetate |
| Show More |