Learn More
Methyl (2RS)-2-Hydroxy-2-phenylacetate (Methyl Mandelate), Mikromol™
Description
Methyl (2RS)-2-Hydroxy-2-phenylacetate (Methyl Mandelate)
Specifications
Specifications
| Chemical Name or Material | Methyl (2RS)-2-Hydroxy-2-phenylacetate (Methyl Mandelate) |
| CAS | 4358-87-6 |
| Analyte or Component Names | Methyl (2RS)-2-Hydroxy-2-phenylacetate (Methyl Mandelate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H10 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Homatropine Methylbromide Imp. F (EP), Methyl (2RS)-2-Hydroxy-2-phenylacetate, Methyl Mandelate, Homatropine Methylbromide Impurity F |
| InChI Formula | InChI=1S/C9H10O3/c1-12-9(11)8(10)7-5-3-2-4-6-7/h2-6,8,10H,1H3 |
| SMILES | COC(=O)C(O)c1ccccc1 |
| IUPAC Name | methyl 2-hydroxy-2-phenylacetate |
| Show More |