missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (2S)-2-(6-Methoxynaphthalen-2-yl)propanoate (Naproxen Methyl Ester), Mikromol™
Description
Methyl (2S)-2-(6-Methoxynaphthalen-2-yl)propanoate (Naproxen Methyl Ester)
Specifications
Specifications
| Chemical Name or Material | Methyl (2S)-2-(6-Methoxynaphthalen-2-yl)propanoate (Naproxen Methyl Ester) |
| CAS | 26159-35-3 |
| Analyte or Component Names | Methyl (2S)-2-(6-Methoxynaphthalen-2-yl)propanoate (Naproxen Methyl Ester) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H16 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methyl (2S)-2-(6-Methoxynaphthalen-2-yl)propanoate, Naproxen Imp. E (EP), Naproxen Methyl Ester, Naproxen Impurity E |
| InChI Formula | InChI=1S/C15H16O3/c1-10(15(16)18-3)11-4-5-13-9-14(17-2)7-6-12(13)8-11/h4-10H,1-3H3/t10-/m0/s1 |
| SMILES | COC(=O)[C@@H](C)c1ccc2cc(OC)ccc2c1 |
| IUPAC Name | methyl (2S)-2-(6-methoxynaphthalen-2-yl)propanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?