Learn More
Methyl 3-[[(2RS)-2-(Ethylamino)propanoyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride (Ethylarticaine Hydrochloride), Mikromol™
Product Code. 17922046
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922046 100 mg 100mg
17951566 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17922046 Supplier Mikromol Supplier No. MM0369.040100
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 3-[[(2RS)-2-(Ethylamino)propanoyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride (Ethylarticaine Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ethylarticaine Hydrochloride
CAS 2602451-79-4
Analyte or Component Names Ethylarticaine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H18 N2 O3 S . Cl H
Certifications/Compliance ISO 17025
Synonym Articaine Hydrochloride Imp. D (EP), Articaine Imp. D (EP), Methyl 3-[[(2RS)-2-(Ethylamino)propanoyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride, Articaine Hydrochloride Imp. D (EP) as Hydrochloride, Ethylarticaine Hydrochloride, Articaine Hydrochloride Impurity D as Hydrochloride, Articaine Impurity D as Hydrochloride
InChI Formula InChI=1S/C19H21ClN2.ClH/c1-21(2)12-5-13-22-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)22;/h3-4,6-11,14H,5,12-13H2,1-2H3;1H
SMILES Cl.CCNC(C)C(=O)Nc1c(C)csc1C(=O)OC
IUPAC Name methyl 3-[2-(ethylamino)propanoylamino]-4-methylthiophene-2-carboxylate;hydrochloride
Molecular Weight (g/mol) 306.81
Quantity 100 mg
Formula Weight 306.0805 g/mol