Learn More
Methyl 3-[[(2RS)-2-(Ethylamino)propanoyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride (Ethylarticaine Hydrochloride), Mikromol™
Description
Methyl 3-[[(2RS)-2-(Ethylamino)propanoyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride (Ethylarticaine Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | Ethylarticaine Hydrochloride |
| CAS | 2602451-79-4 |
| Analyte or Component Names | Ethylarticaine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H18 N2 O3 S . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Articaine Hydrochloride Imp. D (EP), Articaine Imp. D (EP), Methyl 3-[[(2RS)-2-(Ethylamino)propanoyl]amino]-4-methylthiophene-2-carboxylate Hydrochloride, Articaine Hydrochloride Imp. D (EP) as Hydrochloride, Ethylarticaine Hydrochloride, Articaine Hydrochloride Impurity D as Hydrochloride, Articaine Impurity D as Hydrochloride |
| InChI Formula | InChI=1S/C19H21ClN2.ClH/c1-21(2)12-5-13-22-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)22;/h3-4,6-11,14H,5,12-13H2,1-2H3;1H |
| SMILES | Cl.CCNC(C)C(=O)Nc1c(C)csc1C(=O)OC |
| IUPAC Name | methyl 3-[2-(ethylamino)propanoylamino]-4-methylthiophene-2-carboxylate;hydrochloride |
| Show More |