missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-bromophenyl)propionate, 97%, Thermo Scientific Chemicals

Catalog Number 15448108
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
75567-84-9
C10H11BrO2
243.1
MFCD09953150
FKPYNBFWCSTPOT-UHFFFAOYSA-N
methyl 3-4-bromophenyl propanoate, methyl 3-4-bromophenyl propionate, 3-4-bromophenyl propanoic acid methyl ester, benzenepropanoic acid, 4-bromo-, methyl ester, 3-4-bromophenyl propionic acid methyl ester, acmc-209oyw, 3-4-bromophenyl propionate, ksc498a6l, methyl3-4-bromophenyl propionate, 3-4-bromophenyl-propionic acid methyl ester
11447883
methyl 3-(4-bromophenyl)propanoate
COC(=O)CCC1=CC=C(C=C1)Br
This item is not returnable. View return policy
75567-84-9
C10H11BrO2
243.1
MFCD09953150
FKPYNBFWCSTPOT-UHFFFAOYSA-N
methyl 3-4-bromophenyl propanoate, methyl 3-4-bromophenyl propionate, 3-4-bromophenyl propanoic acid methyl ester, benzenepropanoic acid, 4-bromo-, methyl ester, 3-4-bromophenyl propionic acid methyl ester, acmc-209oyw, 3-4-bromophenyl propionate, ksc498a6l, methyl3-4-bromophenyl propionate, 3-4-bromophenyl-propionic acid methyl ester
11447883
methyl 3-(4-bromophenyl)propanoate
COC(=O)CCC1=CC=C(C=C1)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Quantity 5 g
Formula Weight 243.1
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-(4-bromophenyl)propionate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'