missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-bromo-2-fluorobenzoate, 98%, Thermo Scientific Chemicals

Catalog Number 15447707
196.67 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
206551-41-9
C8H6BrFO2
233.036
MFCD09261255
ZWOFHFOFKBYRHV-UHFFFAOYSA-N
3-bromo-2-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-2-fluoro-, methyl ester, pubchem21940, acmc-209fct, methyl bromofluorobenzoate, methyl3-bromo-2-fluorobenzoate, 3-bromo-2-fluoro-benzoic acid methyl ester
21584505
methyl 3-bromo-2-fluorobenzoate
COC(=O)C1=C(C(=CC=C1)Br)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
206551-41-9
C8H6BrFO2
233.036
MFCD09261255
ZWOFHFOFKBYRHV-UHFFFAOYSA-N
3-bromo-2-fluorobenzoic acid methyl ester, benzoic acid, 3-bromo-2-fluoro-, methyl ester, pubchem21940, acmc-209fct, methyl bromofluorobenzoate, methyl3-bromo-2-fluorobenzoate, 3-bromo-2-fluoro-benzoic acid methyl ester
21584505
methyl 3-bromo-2-fluorobenzoate
COC(=O)C1=C(C(=CC=C1)Br)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 35°C to 37°C
Color White to Cream
Quantity 5 g
Formula Weight 233.04
Percent Purity 98%
Physical Form Fused Solid
Chemical Name or Material Methyl 3-bromo-2-fluorobenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'