missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Bromo-2-fluorophenylacetate, 96%, Thermo Scientific Chemicals

Catalog Number p-7054446
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1427451-43-1
C9H8BrFO2
247.063
ALPXGXOFMZOFAG-UHFFFAOYSA-N
methyl 3-bromo-2-fluorophenylacetate, methyl 2-3-bromo-2-fluorophenyl acetate
100912177
methyl 2-(3-bromo-2-fluorophenyl)acetate
COC(=O)CC1=C(C(=CC=C1)Br)F
This item is not returnable. View return policy
1427451-43-1
C9H8BrFO2
247.063
ALPXGXOFMZOFAG-UHFFFAOYSA-N
methyl 3-bromo-2-fluorophenylacetate, methyl 2-3-bromo-2-fluorophenyl acetate
100912177
methyl 2-(3-bromo-2-fluorophenyl)acetate
COC(=O)CC1=C(C(=CC=C1)Br)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 247.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Methyl 3-Bromo-2-fluorophenylacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'