Learn More
Methyl 3-Cyanoropionate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 3-Cyanoropionate |
| CAS | 4107-62-4 |
| Molecular Formula | C5H7NO2 |
| Quantity | 2.5 g |
| Synonym | 3-Carbomethoxypropionitrile, 3-Cyanopropionic acid methyl ester, Methyl 3-Cyanopropanoate, Methyl beta-cyanopropionate, NSC 71943 |
| InChI Formula | InChI=1S/C5H7NO2/c1-8-5(7)3-2-4-6/h2-3H2,1H3 |
| SMILES | COC(=O)CCC#N |
| IUPAC Name | methyl 3-cyanopropanoate |
| Molecular Weight (g/mol) | 113.11 |
| Formula Weight | 113.0477 |