missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-fluoro-4-nitrobenzoate, 97%, Thermo Scientific Chemicals

Catalog Number p-7055962
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
185629-31-6
C8H6FNO4
199.137
MFCD08444027
FKMZNQQOPCCUTD-UHFFFAOYSA-N
592761
methyl 3-fluoro-4-nitrobenzoate
COC(=O)C1=CC(=C(C=C1)[N+](=O)[O-])F
This item is not returnable. View return policy
185629-31-6
C8H6FNO4
199.137
MFCD08444027
FKMZNQQOPCCUTD-UHFFFAOYSA-N
592761
methyl 3-fluoro-4-nitrobenzoate
COC(=O)C1=CC(=C(C=C1)[N+](=O)[O-])F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 5 g
Beilstein 10657755
Synonym methyl 3-fluoro-4-nitrobenzenecarboxylate, methyl3-fluoro-4-nitrobenzenecarboxylate, 3-fluoro-4-nitrobenzoic acid methyl ester, benzoic acid, 3-fluoro-4-nitro-, methyl ester, 2-fluoro-4-methoxycarbonyl nitrobenzene, 3-fluoro-4-nitro-benzoic acid methyl ester, pubchem15965, acmc-209en1, ksc495e5t, methyl 3-fluro-4nitrobenzoate
Formula Weight 199.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Methyl 3-fluoro-4-nitrobenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'