missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(N-Benzylamino)propionate, TRC

Product Code. 30471929
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471929 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471929 Supplier Toronto Research Chemicals Supplier No. TRCM28899510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 3-(N-Benzylamine)propionate
CAS 23574-01-8
Molecular Formula C11 H15 N O2
Quantity 10 g
Synonym β-Alanine, N-(phenylmethyl)-, methyl ester, β-Alanine, N-benzyl-, methyl ester (8CI), Methyl 3-(benzylamino)propanoate, Methyl 3-(benzylamino)propionate, Methyl N-(benzyl)-3-aminopropionate, Methyl N-benzyl-β-alaninate, N-Benzyl-β-alanine methyl ester, 3-(Benzylamino)propanoic acid methyl ester
InChI Formula InChI=1S/C11H15NO2/c1-14-11(13)7-8-12-9-10-5-3-2-4-6-10/h2-6,12H,7-9H2,1H3
SMILES COC(=O)CCNCc1ccccc1
IUPAC Name methyl 3-(benzylamino)propanoate
Molecular Weight (g/mol) 193.24
Formula Weight 193.1103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.