Learn More
Methyl 3-O-Benzyl-D-glycerate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 3-O-Benzyl-D-glycerate |
| CAS | 209907-54-0 |
| Molecular Formula | C11 H14 O4 |
| Quantity | 250 mg |
| Synonym | Propanoic acid, 2-hydroxy-3-(phenylmethoxy)-, methyl ester, (2R)-, (R)-3-[(Benzyl)oxy]-2-hydroxypropionic acid methyl ester, (R)-Methyl 3-(benzyloxy)-2-hydroxypropanoate, Methyl (R)-3-benzyloxy-2-hydroxypropionate, (2R)-3-(Benzyloxy)-2-hydroxypropanoic acid methyl ester |
| InChI Formula | InChI=1S/C11H14O4/c1-14-11(13)10(12)8-15-7-9-5-3-2-4-6-9/h2-6,10,12H,7-8H2,1H3/t10-/m1/s1 |
| SMILES | COC(=O)[C@H](O)COCc1ccccc1 |
| IUPAC Name | methyl (2R)-2-hydroxy-3-phenylmethoxypropanoate |
| Molecular Weight (g/mol) | 210.23 |
| Formula Weight | 210.0892 |