Learn More
Methyl 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate, 97%, Thermo Scientific™
Product Code. 15458585
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458585 5 g 5g
15468585 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15458585 Supplier Thermo Scientific Alfa Aesar Supplier No. H33685.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 202195-67-3
Molecular Formula C10H9NO4
Molecular Weight (g/mol) 207.19
MDL Number MFCD03036646
InChI Key WPGYGHFVLMZWHI-UHFFFAOYSA-N
Synonym methyl 3-oxo-3,4-dihydro-2h-1,4-benzoxazine-6-carboxylate, methyl 3-oxo-3,4-dihydro-2h-benzo b 1,4 oxazine-6-carboxylate, methyl 3,4-dihydro-3-oxo-2h-benzo b 1,4 oxazine-6-carboxylate, 2h-1,4-benzoxazine-6-carboxylic acid, 3,4-dihydro-3-oxo-, methyl ester, 3-oxo-3,4-dihydro-2h-benzo 1,4 oxazine-6-carboxylic acid methyl ester, methyl 3-oxo-2,4-dihydro-1,4-benzoxazine-6-carboxylate, methyl 1,4-benzoxazine-3-one-6-carboxylate, 3,4-dihydro-6-methoxycarbonyl-3-oxo-2h-1,4-benzoxazine, methyl-3-oxo-3,4-dihydro-2h-1,4-benzoxazin-6-carboxylate, methyl 3,4-dihydro-3-oxo-2h-1,4-benzoxazine-6-carboxylate
PubChem CID 2974054
IUPAC Name methyl 3-oxo-4H-1,4-benzoxazine-6-carboxylate
SMILES COC(=O)C1=CC2=C(OCC(=O)N2)C=C1
Specifications
Melting Point 182°C to 184°C
Quantity 5 g
Formula Weight 207.19
Percent Purity 97%
Chemical Name or Material Methyl 3-oxo-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate