Learn More
Methyl 3-Oxoheptanoate, TRC
Specifications
Specifications
| Chemical Name or Material | Methyl 3-Oxoheptanoate |
| CAS | 39815-78-6 |
| Molecular Formula | C8H14O3 |
| Quantity | 25 g |
| Synonym | 3-oxo-Heptanoic Acid Methyl Ester, 3-Oxoheptanoic Acid Methyl Ester, Methyl Pentanoylacetate |
| InChI Formula | InChI=1S/C8H14O3/c1-3-4-5-7(9)6-8(10)11-2/h3-6H2,1-2H3 |
| SMILES | CCCCC(=O)CC(=O)OC |
| IUPAC Name | methyl 3-oxoheptanoate |
| Molecular Weight (g/mol) | 158.19 |
| Formula Weight | 158.0943 |