Learn More
Methyl 4-aminobutyrate hydrochloride, 99%, Thermo Scientific Chemicals
Catalog Number p-4813551
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
13031-60-2
C5H12ClNO2
153.61
MFCD00043270
WPGPRLVPWACBHW-UHFFFAOYSA-N
methyl 4-aminobutanoate hydrochloride, methyl 4-aminobutyrate hydrochloride, methyl 4-aminobutanoate hcl, methyl 4-aminobutyrate hcl, h-gamma-abu-ome.hcl, 4-aminobutyric acid methyl ester hydrochloride, 4-aminobutyric acid methyl ester, hydrochloride, 4-amino-n-butyric acid methyl ester hydrochloride, butanoic acid, 4-amino-, methyl ester, hydrochloride, 4-amino-butyric acid methyl ester hydrochloride
2756383
methyl 4-aminobutanoate;hydrochloride
[Cl-].COC(=O)CCC[NH3+]
13031-60-2
C5H12ClNO2
153.61
MFCD00043270
WPGPRLVPWACBHW-UHFFFAOYSA-N
methyl 4-aminobutanoate hydrochloride, methyl 4-aminobutyrate hydrochloride, methyl 4-aminobutanoate hcl, methyl 4-aminobutyrate hcl, h-gamma-abu-ome.hcl, 4-aminobutyric acid methyl ester hydrochloride, 4-aminobutyric acid methyl ester, hydrochloride, 4-amino-n-butyric acid methyl ester hydrochloride, butanoic acid, 4-amino-, methyl ester, hydrochloride, 4-amino-butyric acid methyl ester hydrochloride
2756383
methyl 4-aminobutanoate;hydrochloride
[Cl-].COC(=O)CCC[NH3+]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 118°C to 119°C
Assay Percent Range 98.5 to 101.5% (Argentometry)
Packaging Glass Bottle
Formula Weight 153.61
Percent Purity 99%
Quantity 25 g
Chemical Name or Material Methyl 4-aminobutyrate hydrochloride
compliance-icons
Product Identifier
  • Methyl 4-aminobutyrate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.