missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Hydroxybenzoate-2,3,5,6-d4, CDN
missing translation for 'orderingAttributeHoverText'
Quantity:
0.05 g
0.1 g
missing translation for 'unitSize'
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Methyl Paraben-d4 |
| CAS | 362049-51-2 |
| Molecular Formula | C8 2H4 H4 O3 |
| Quantity | 0.1 g |
| Synonym | Benzoic-2,3,5,6-d4 acid, 4-hydroxy-, methyl ester (9 CI), Methyl 4-hydroxybenzoate-2,3,5,6-D4, Methyl 4-hydroxybenzoate-d4, Methyl parahydroxybenzoate-d4, 4-Hydroxybenzoic-2,3,5,6-d4 acid methyl ester, Methylparaben-d4 |
| InChI Formula | InChI=1 S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9 H,1H3/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)OC)c([2 H])c([2 H])c1O |
| IUPAC Name | methyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate |
| Molecular Weight (g/mol) | 156.17 |
| Formula Weight | 156.0725 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction